Order Entry
Austria
ContactUsLinkComponent
XELA UHPLC Columns, Biphenyl
XELA UHPLC Columns, Biphenyl
Supplier:  ISERA
Certificates
XELA UHPLC Columns, Biphenyl
Supplier:  ISERA

Specifications

  • Catalog No:
  • ISERA211-N11X00
  • ISERA211-N11A00
  • ISERA211-N11U00
  • ISERA211-N11V00
  • ISERA211-N11K00
  • ISERA211-N11M00
  • ISERA211-N11G00
  • ISERA211-N11H00
  • ISERA211-N11D00
  • ISERA211-N11F00
  • ISERA211-N11Z00
  • ISERA211-N11B00
  • Phase name:
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • Application:
  • Suitable for aromatics and medium-polar analytes
  • Suitable for aromatics and medium-polar analytes
  • Suitable for aromatics and medium-polar analytes
  • Suitable for aromatics and medium-polar analytes
  • Suitable for aromatics and medium-polar analytes
  • Suitable for aromatics and medium-polar analytes
  • Suitable for aromatics and medium-polar analytes
  • Suitable for aromatics and medium-polar analytes
  • Suitable for aromatics and medium-polar analytes
  • Suitable for aromatics and medium-polar analytes
  • Suitable for aromatics and medium-polar analytes
  • Suitable for aromatics and medium-polar analytes
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Max. pressure:
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • pH range:
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • USP code:
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • Storage:
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • Shipping conditions:
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.

Specifications

Frequently Bought Together