Order Entry
Austria
ContactUsLinkComponent
XELA UHPLC Columns, HILIC-Diol
XELA UHPLC Columns, HILIC-Diol
Supplier:  ISERA
Certificates
XELA UHPLC Columns, HILIC-Diol
Supplier:  ISERA

Specifications

  • Catalog No:
  • ISERA211-G11U00
  • ISERA211-G31Z00
  • ISERA211-G11V00
  • ISERA211-G31U00
  • ISERA211-G11M00
  • ISERA211-G31V00
  • ISERA211-G31M00
  • ISERA211-G11G00
  • ISERA211-G11H00
  • ISERA211-G31G00
  • ISERA211-G11B00
  • ISERA211-G31H00
  • ISERA211-G11F00
  • ISERA211-G31B00
  • ISERA211-G31F00
  • ISERA211-G11A00
  • ISERA211-G11Z00
  • ISERA211-G31A00
  • Phase name:
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Max. pressure:
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • pH range:
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • USP code:
  • L20
  • L20
  • L20
  • L20
  • L20
  • L20
  • L20
  • L20
  • L20
  • L20
  • L20
  • L20
  • L20
  • L20
  • L20
  • L20
  • L20
  • L20
  • Storage:
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • Shipping conditions:
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.

Specifications

Frequently Bought Together