Order Entry
Austria
ContactUsLinkComponent
SERAspher HPLC Columns, BioRP
SERAspher HPLC Columns, BioRP
Supplier:  ISERA
Certificates
SERAspher HPLC Columns, BioRP
Supplier:  ISERA

Specifications

  • Catalog No:
  • ISERA1P1-A25D00
  • ISERA1P1-A25E00
  • ISERA1P1-A25B00
  • ISERA1P1-A25C00
  • ISERA1P1-A25A00
  • ISERA1P1-A25A0B
  • ISERA1P1-A25U00
  • ISERA1P1-A25V00
  • ISERA1P1-A25S00
  • ISERA1P1-A25T00
  • ISERA1P1-A25Q00
  • ISERA1P1-A25R00
  • ISERA1P1-A25N00
  • ISERA1P1-A25P00
  • ISERA1P1-A25L00
  • ISERA1P1-A25M00
  • ISERA1P2-A25L00
  • ISERA1P2-A25L0B
  • ISERA1P1-A25Y0B
  • ISERA1P1-A25Z00
  • ISERA1P1-A25X00
  • ISERA1P1-A25Y00
  • ISERA1P1-A25J00
  • ISERA1P1-A25W00
  • ISERA1P1-A25K00
  • ISERA1P1-A25G00
  • ISERA1P1-A25H00
  • Phase name:
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • SERAspher
  • Application:
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Separation and purification of peptides (biocompatible surface-coated stainless steel column hardware for models ISERA1P1-A25A0B, ISERA1P1-A25Y0B, and ISERA1P2-A25L0B only)
  • Max. pressure:
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • 200 bar
  • pH range:
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • 1 - 14
  • USP code:
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • Storage:
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • Shipping conditions:
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.

Specifications

Frequently Bought Together