Order Entry
Austria
ContactUsLinkComponent
ISAspher HPLC Columns, C8 Select
ISAspher HPLC Columns, C8 Select
Supplier:  ISERA
Certificates
ISAspher HPLC Columns, C8 Select
Supplier:  ISERA

Specifications

  • Catalog No:
  • ISERA111-R25W00
  • ISERA111-R25X00
  • ISERA111-R25U00
  • ISERA111-R25V00
  • ISERA111-R25S00
  • ISERA111-R25T00
  • ISERA111-R25R00
  • ISERA111-R25P00
  • ISERA111-R25Q00
  • ISERA111-R25M00
  • ISERA111-R25N00
  • ISERA111-R25K00
  • ISERA111-R25L00
  • ISERA111-R25H00
  • ISERA111-R25J00
  • ISERA111-R25F00
  • ISERA111-R25G00
  • ISERA111-R25E00
  • ISERA111-R25C00
  • ISERA111-R25D00
  • ISERA111-R25A00
  • ISERA111-R25Y00
  • ISERA111-R25B00
  • ISERA111-R25Z00
  • Phase name:
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • Carbon load:
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • 10%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Max. pressure:
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • pH range:
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • 2,5 - 7,5
  • Surface area:
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • USP code:
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • Storage:
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • Shipping conditions:
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.

Specifications

Frequently Bought Together