ISAspher Reversed Phase HPLC Columns, Amino
: ISERA
HPLC column of the ISAspher line guarantee the highest possible reliability in a wide range of applications.
- The production of the silica used as well as the functionalisation are carried out according to highly developed and validated process
- Reliable and proven
- Flexibility and robustness for every HPLC application
This ensure unchanged properties of the stationary phases in terms of selectivity and consistent chemical properties.
Direct scale-up e.g. starting from a particle size of 5 µm to semi-preparative or preparative applications (up to 45 µm particle size) can be easily implemented. Of course, the same applies for a scale-down, e.g. to a 3 µm particle size, if a speed-optimised analytical method has to be established.
: Individual test certificate is enclosed with each column.
: Columns are delivered in solvent which is suitable for reversed phase / HILIC chromatography.
- Catalog No:
- ISERA112-J23F00
- ISERA112-J23C00
- ISERA112-J23E00
- ISERA112-J23A00
- ISERA112-J23B00
- ISERA112-J23F30
- ISERA112-J23G30
- ISERA112-J23C30
- ISERA112-J23E30
- ISERA112-J23B30
- ISERA112-J25G00
- ISERA112-J23A30
- ISERA112-J25E00
- ISERA112-J25F00
- ISERA112-J25B00
- ISERA112-J25C00
- ISERA112-J25G30
- ISERA112-J25A00
- ISERA112-J25E30
- ISERA112-J25F30
- ISERA111-J29Z00
- ISERA112-J29G00
- ISERA111-J29X00
- ISERA112-J25C30
- ISERA111-J29Y00
- ISERA111-J29V00
- ISERA112-J25A30
- ISERA112-J29E00
- ISERA112-J29F00
- ISERA112-J25B30
- ISERA111-J29W00
- ISERA111-J29T00
- ISERA111-J29U00
- ISERA111-J29S00
- ISERA111-J29Q00
- ISERA111-J29R00
- ISERA111-J29N00
- ISERA111-J29P00
- ISERA111-J29L00
- ISERA111-J29M00
- ISERA111-J29J00
- ISERA111-J29K00
- ISERA111-J29G00
- ISERA111-J29H00
- ISERA111-J29F00
- ISERA111-J29D00
- ISERA111-J29E00
- ISERA111-J29B00
- ISERA111-J29C00
- ISERA111-J25Z00
- ISERA111-J29A00
- ISERA111-J23N00
- ISERA111-J25X00
- ISERA111-J25Y00
- ISERA111-J23P00
- ISERA111-J23L00
- ISERA111-J25W00
- ISERA111-J23M00
- ISERA111-J23J00
- ISERA111-J23K00
- ISERA111-J23G00
- ISERA111-J23H00
- ISERA111-J23E00
- ISERA111-J23F00
- ISERA111-J25U00
- ISERA111-J25V00
- ISERA111-J25S00
- ISERA111-J25T00
- ISERA111-J25Q00
- ISERA111-J25R00
- ISERA111-J25N00
- ISERA111-J23D00
- ISERA111-J25P00
- ISERA111-J23B00
- ISERA111-J25L00
- ISERA111-J23C00
- ISERA111-J25M00
- ISERA111-J23A00
- ISERA111-J25K00
- ISERA111-J25H00
- ISERA111-J25J00
- ISERA111-J25F00
- ISERA111-J25G00
- ISERA111-J25D00
- ISERA111-J25E00
- ISERA111-J25B00
- ISERA111-J25C00
- ISERA111-J25A00
- ISERA111-J23Y00
- ISERA111-J23Z00
- ISERA111-J23W00
- ISERA111-J23X00
- ISERA111-J23U00
- ISERA111-J23V00
- ISERA111-J23S00
- ISERA111-J23T00
- ISERA111-J23Q00
- ISERA111-J23R00
- ISERA112-J29B00
- ISERA112-J29C00
- ISERA112-J29G30
- ISERA112-J29A00
- ISERA112-J29E30
- ISERA112-J29F30
- ISERA112-J29B30
- ISERA112-J29C30
- ISERA112-J23G00
- ISERA112-J29A30
- Phase name:
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- ISAspher
- Carbon load:
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- pH range:
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- 3 - 7
- Surface area:
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- USP code:
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- L8
- Storage:
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- RT
- Shipping conditions:
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
- Shipped under testing eluent.
Guard column holders
Frequently Bought Together