VWR® Culture Tubes With Screw Thread
Reusable culture tubes with screw thread made from clear borosilicate glass.
- High resistance against chemicals
- High thermal resistance
- With white marking field
: Screw caps have to be ordered separately.
Reusable culture tubes with screw thread made from clear borosilicate glass.