Orders LinkContactUsLinkComponent
EPREP Labels, M710
Supplier:   Brady
EPREP Labels, M710
Supplier:  Brady
Certificates
EPREP Labels, M710
EPREP Labels, M710
21 Options Available Below
Product Details & Documents
Thermal transfer printable raised panel labels. Designed to make legend plate creation quick, less expensive, and hassle-free.
  • Aggressive adhesive and solvent resistance qualities are a great replacement for engraved tags
  • Service temperature of −40 to 100 °C
  • Gloss finish
Use for electrical components, electrical cabinets, push buttons, patch panels, and any other applications required. This material can also be used for rating and serial plates using alphanumerics that require name plate quality.
Product Family Options
Product Information
  • Model
    Material
    Colour
    Dimensions
    Packed
    Availability
    Price
  • M7-1-7593-SL
    Polyester
    Silver
    27,00W×7,95H mm
    250/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 67,50 €
    67,50 €
    1 * 1 Rolle
       67,50 €
  • M7-1-7593-WT
    Polyester
    White
    27,00W×7,95H mm
    250/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 67,50 €
    67,50 €
    1 * 1 Rolle
       67,50 €
  • M7-2-7593-SL
    Polyester
    Silver
    27,00W×12,45H mm
    250/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 88,30 €
    88,30 €
    1 * 1 Rolle
       88,30 €
  • M7-2-7593-WT
    Polyester
    White
    27,00W×12,45H mm
    250/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 88,30 €
    88,30 €
    1 * 1 Rolle
       88,30 €
  • M7-3-7593-SL
    Polyester
    Silver
    27,00W×15,00H mm
    150/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 67,70 €
    67,70 €
    1 * 1 Rolle
       67,70 €
  • M7-3-7593-WT
    Polyester
    White
    27,00W×15,00H mm
    150/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 67,70 €
    67,70 €
    1 * 1 Rolle
       67,70 €
  • M7-4-7593-SL
    Polyester
    Silver
    27,00W×18,00H mm
    150/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 77,40 €
    77,40 €
    1 * 1 Rolle
       77,40 €
  • M7-4-7593-WT
    Polyester
    White
    27,00W×18,00H mm
    150/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 77,40 €
    77,40 €
    1 * 1 Rolle
       77,40 €
  • M7-5-7593-GN
    Polyester
    Green
    30,00W×40,00H mm
    100/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 83,30 €
    83,30 €
    1 * 1 Rolle
       83,30 €
  • M7-5-7593-RD
    Polyester
    Red
    30,00W×40,00H mm
    100/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 83,30 €
    83,30 €
    1 * 1 Rolle
       83,30 €
  • M7-5-7593-SL
    Polyester
    Silver
    30,00W×40,00H mm
    100/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 83,30 €
    83,30 €
    1 * 1 Rolle
       83,30 €
  • M7-5-7593-WT
    Polyester
    White
    30,00W×40,00H mm
    100/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 83,30 €
    83,30 €
    1 * 1 Rolle
       83,30 €
  • M7-5-7593-YL
    Polyester
    Yellow
    30,00W×40,00H mm
    100/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 83,30 €
    83,30 €
    1 * 1 Rolle
       83,30 €
  • M7-7-7593-SL
    Polyester
    Silver
    35,00W×18,00H mm
    100/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 71,80 €
    71,80 €
    1 * 1 Rolle
       71,80 €
  • M7-7-7593-WT
    Polyester
    White
    35,00W×18,00H mm
    100/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 71,80 €
    71,80 €
    1 * 1 Rolle
       71,80 €
  • M7-7-7593-YL
    Polyester
    Yellow
    35,00W×18,00H mm
    100/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 71,80 €
    71,80 €
    1 * 1 Rolle
       71,80 €
  • M7-6-7593-BK
    Polyester
    Black
    45,00W×15,00H mm
    100/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 91,40 €
    91,40 €
    1 * 1 Rolle
       91,40 €
  • M7-6-7593-RD
    Polyester
    Red
    45,00W×15,00H mm
    100/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 91,40 €
    91,40 €
    1 * 1 Rolle
       91,40 €
  • M7-6-7593-YL
    Polyester
    Yellow
    45,00W×15,00H mm
    100/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 91,40 €
    91,40 €
    1 * 1 Rolle
       91,40 €
  • M7-8-7593-SL
    Polyester
    Silver
    48,00W×19,00H mm
    125/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 147,00 €
    147,00 €
    1 * 1 Rolle
       147,00 €
  • M7-8-7593-WT
    Polyester
    White
    48,00W×19,00H mm
    125/roll
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 147,00 €
    147,00 €
    1 * 1 Rolle
       147,00 €