Destructible Tamper Evident Matte Vinyl Labels for M710 and BMP71
Supplier: Brady
Certificates
About this item
Tamper-resistant vinyl (B-351) is designed to fracture easily in order to show signs of product tampering and to prevent on-piece label removal.
- High performance white matte vinyl with permanent adhesive
- Ideal for applications that require resistance to product tampering
- Use caution when removing from liner as material is fragile
- Service temperature of −40 to +82 °C (−40 to +180 °F)
Brady Authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6200.
Frequently Bought Together