Self-Laminating Vinyl Wrap Around Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Supplier: Brady
Certificates
About this item
Self-laminating vinyl (B-427) is an excellent material for wire and cable identification. This material has good clarity and conformability, and is self-extinguishing.
- Rotate to identify labels from any angle
- Simply wrap and apply; no adhesive residue or label parts will cling to wires
- Spins freely without anchors so these labels won’t damage Cat 5/6 and fiber optic cables
- Durable, permanent acrylic adhesive has outstanding water and oil resistance
- Service temperature of −40 to +70 °C (−40 to +158 °F)
- Matte finish
Ideal for data and telecommunications labelling, electrical labelling, laboratory labelling, and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R4300 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Certifications: UL recognised to UL969 labelling and marking standard when printed with the Brady series R4300 and R6200 ribbons. See UL files MH17154 for specific details. Brady customers should specify at purchase if they require UL recognised B-427 labels. Brady sells both UL recognised B-427 labels and non-approved labels depending on customer requirements.
Frequently Bought Together