Orders LinkContactUsLinkComponent
Self-laminating Polyester Labels for M710
Self-laminating Polyester Labels for M710
Supplier:   Brady
Self-laminating Polyester Labels for M710
Supplier:  Brady
Certificates
Self-laminating Polyester Labels for M710
Self-laminating Polyester Labels for M710
9 Options Available Below
Product Details & Documents
Self-laminating polyester labels for M710 printer.
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance, and high/low temperature performance
Ideal for laboratory labelling applications.
Product Family Options
Product Information
  • Type
    Model
    Material
    Adhesive
    Colour
    Printer model
    Dimensions
    Packed
    Availability
    Price
  • BRDY311988
    Labels, rectangular
    Labels, rectangular
    B-461
    M7-73-461
    Polyester
    Permanent acrylic
    White/Transparent
    M710
    9,53W×45,72H mm
    250/box
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 275,00 €
    275,00 €
    1 * 1 Rolle
       275,00 €
  • BRDY311973
    Labels, rectangular
    Labels, rectangular
    B-461
    M7-100-461
    Polyester
    Permanent acrylic
    White/Transparent
    M710
    15,24W×38,10H mm
    500/box
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 200,00 €
    200,00 €
    1 * 1 Rolle
       200,00 €
  • BRDY311989
    Labels, rectangular
    Labels, rectangular
    B-461
    M7-76-461
    Polyester
    Permanent acrylic
    White/Transparent
    M710
    15,24W×41,28H mm
    500/box
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 127,00 €
    127,00 €
    1 * 1 Rolle
       127,00 €
  • BRDY311979
    Labels, rectangular
    Labels, rectangular
    B-461
    M7-119-461
    Polyester
    Permanent acrylic
    White/Transparent
    M710
    15,24W×48,26H mm
    500/box
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 253,00 €
    253,00 €
    1 * 1 Rolle
       253,00 €
  • BRDY311992
    Labels, rectangular
    Labels, rectangular
    B-461
    M7-86-461
    Polyester
    Permanent acrylic
    White/Transparent
    M710
    15,24W×66,68H mm
    100/box
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 69,90 €
    69,90 €
    1 * 1 Rolle
       69,90 €
  • BRDY311975
    Labels, rectangular
    Labels, rectangular
    B-461
    M7-106-461
    Polyester
    Permanent acrylic
    White/Transparent
    M710
    25,40W×44,45H mm
    250/box
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 195,00 €
    195,00 €
    1 * 1 Rolle
       195,00 €
  • BRDY311976
    Labels, rectangular
    Labels, rectangular
    B-461
    M7-115-461
    Polyester
    Permanent acrylic
    White/Transparent
    M710
    25,40W×66,68H mm
    100/box
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 117,00 €
    117,00 €
    1 * 1 Rolle
       117,00 €
  • BRDY311977
    Labels, rectangular and round
    Labels, rectangular and round
    B-116
    M7-116-461
    Polyester
    Permanent acrylic
    White/Transparent
    M710
    15,24W×41,28H×12,70Ø mm
    150 sets/box
    Log in for availability
    Log in for availability
    1 * 150 SET
    1 * 150 SET - 64,90 €
    64,90 €
    1 * 150 SET
       64,90 €
  • BRDY311974
    Labels, rectangular and round
    Labels, rectangular and round
    B-101
    M7-101-461
    Polyester
    Permanent acrylic
    White/Transparent
    M710
    40,00W×15,00H×9,53Ø m
    250 sets/box
    Log in for availability
    Log in for availability
    1 * 250 SET
    1 * 250 SET - 104,00 €
    104,00 €
    1 * 250 SET
       104,00 €