Electrostatic Dissipative (ESD) Polyimide Labels for M710 and BMP71
Supplier: Brady
Certificates
About this item
Static Dissipative Polyimide (B-717) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labelling.
- High temperature, glossy white polyimide material
- Static dissipative adhesive and static dissipative release liner
- Surface resistivity values in the recommended range for dissipative ESD packaging materials as defined by ANSI/ESD S541-2008
- Service temperature of −70 to 100 °C (−94 to 212 °F)
Ideal for circuit board labelling.
Brady Authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6000.
Certifications: UL recognised. Meets the requirements of ANSI/ESD S541-2008 (between 10⁴ and 10¹ ohms) for dissipative ESD packaging materials. Meets the performance requirements of MIL-STD-202G, Method 215K.
Frequently Bought Together