Aggressive Adhesive Multi-Purpose Nylon Labels B-499 for M710, BMP71 Printers
Lieferant: Brady
Zertifikate
Über diesen Artikel
Nylon cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
- White polyamide coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable
- Recommended for indoor use
- Matte finish
- Service temperature of −40 to +90 °C (−40 to +193 °F)
Labels are ideal for component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, laboratory labelling, and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R4300.
Certifications: UL recognised. CSA accepted. Halogen-free (DIN VDE 0472/part 815). Approvals are for white material only.
Frequently Bought Together