Ultra Aggressive Adhesive Multi-Purpose Polyester Labels for M710 and BMP71
: Brady
ToughBond® ultra aggressive polyester (B-483) is perfect for labelling products and general ID and can be applied to powder coated, smooth, textured, or irregular surfaces.
- Glossy white polyester with permanent rubber based adhesive is thermal transfer printable
- Material is designed for high adhesion to textured metals and low surface energy plastics
- Service temperature of −40 to +120 °C (−40 to +248 °F)
Labels are ideal for barcode labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, inventory and inspection labelling, and floor marking applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part numbers M7-R6000 or M7-R4400-WT.
: UL recognised. CSA accepted.
Frequently Bought Together