Graphite, powder, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7782-42-5
:
Formula:C
MDL Number:MFCD00144065
Merck Index:12,04560
Melting Pt:3827 °C
Density:2.09…2.23 g/cm³ (20 °C)
:12.01 g/mol
EINECS:231-955-3
Specification Test Results
- Loss on drying (120°C):≤ 2.5 %
- Particle size (< 75 μm):≥ 95 %
- Substances soluble in ethanol:≤ 0.2 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC24488.290 spec_VWRC24488.360 spec_VWRC24488.460
Product specification
Frequently Bought Together