ContactUsLinkComponent
Hydrogen and High Purity Air Generator for GC-FID
Hydrogen and High Purity Air Generator for GC-FID
Catalog # CCDL928276
Supplier:  LUCIDITY
undefined
Hydrogen and High Purity Air Generator for GC-FID
Catalog # CCDL928276
Supplier:  LUCIDITY
Specifications
  • Description:
    Hydrogen and high purity air generator for the Lucidity GC-FID, 266,7×457,2×317,5 mm
  • Pk:
    1
Specifications
About this item
Generator that produces all necessary gasses required to run the Lucidity GC-FID.
  • Hydrogen and high purity air generator for the Lucidity GC-FID
  • Small footprint and economical
  • Self-installable instrument that only requries DI-H₂O
The Lucidity GAS produces hydrogen and high purity air. The economical GAS with its small footprint compliments the Lucidity GC-FID perfectly. The GAS produces all necessary gasses required to run the Lucidity GC-FID. Just add DI-H2O to the self-installable instrument and your lab can have access to gasses required for analysis immediately. The Lucidity GC-FID and GAS communicate with each other, making the perfect and safest pair of instrumentation. With a 24-month warranty the GAS is a great solution for all labs.