Cole-Parmer® CB-200 Block Chiller/Heater
Catalog # TECN36620-17
Supplier: Cole-Parmer
Specifications
- Temperature Range:4...37 °C; uniformity ±0,2 °C (0,36 °F) °C
- No. of blocks:2
- Description:Block chiller/heater, 100 - 230 V, 50/60 Hz
- W×D×H:340×195×225 mm
- Pk:1
Specifications
About this item
Safely maintain biological samples.
- Designed for maintaining biological samples safely between 4 and 37 °C
- Alarm sounds when set temperature is reached and elapsed time ends
- Five-digit LED display can be set for either °C or °F
- Choose from a wide range of interchangeable aluminium block inserts
This benchtop block chiller/heater is ideal for incubating sample tubes at temperatures ranging from 0...40 °C (32...104 °F). The Isotemp® temperature control ensures all contents are kept at an even temperature, whether cold or warm. Integrated countdown/count-up timer with audible alarm allows up to nine days to be programmed and will not start until a stable temperature has been reached.
Use with Dri-Block® aluminium inserts to maintain temperature stability of ±0,2 °C. Unit holds two single-width inserts or one double-width inserts. A dark plastic lid protects light-sensitive reagents as well as helping to maintain the sample temperature.
Delivery: Lid and power cords with plug (US, UK, and EU).