Order Entry
Austria
ContactUsLinkComponent
Sterilin™ Bulk Packed Glass Pipettes
Sterilin™ Bulk Packed Glass Pipettes
Catalog # SLIN7079-2N
Supplier:  Sterilin
undefined
Sterilin™ Bulk Packed Glass Pipettes
Catalog # SLIN7079-2N
Supplier:  Sterilin

Specifications

  • Packed:
    35/pk., 700/cs.
  • Sterile:
    -
  • Capacity:
    2 ml
  • Length:
    290 mm
  • Pk:
    700

Specifications

About this item

Conduct applications with the chemical resistance of Pyrex™ borosilicate glass using Thermo Scientific™ Sterilin™ Bulk Packed Glass Pipettes, in easy to handle shelf packs.

  • Calibrated for delivery with the last drop expelled by blowing
  • Available either plugged or unplugged
  • Easy to read graduations