ContactUsLinkComponent
pH Sensor Rugged, Memosens CPL59E
  662-2461
undefined
pH Sensor Rugged, Memosens CPL59E
  662-2461
  • Type:
    Potentiometric pH glass sensor
  • Description:
    Memosens CPL59E pH sensor rugged, 120 mm; Teflon ring junction, ion trap, 3 M KCl, Ag/AgCl ref. system
  • pH range:
    0 - 14
  • Connection:
    Memosens
  • Range:
    0 - 14 pH, 0...135 °C (application range)
  • Sensor type:
    Basic version, zero point pH 7,0, temperature sensor NTC 30k
  • Reference system:
    Teflon ring junction, ion trap, 3 M KCl, Ag/AgCl
  • Pk:
    1
  • pH:
    0 - 14 pH, 0...135 °C
Potentiometric pH glass sensor for laboratory measurements and random sampling in demanding media. Connectable via inductive, digital connection with Memosens technology.
  • pH sensor for aggressive media
  • Ion trap provides long time stability and stable values in harsh samples with oxidizing agents
  • Large dirt-repellent PTFE junction to avoid soiling
The Memosens 2.0 pH sensor CPL59E shows its performance when other sensors cannot cope with the conditions. This sensor is perfectly suited to harsh conditions such as in the chemical and process industries. Thanks to the ion trap, it offers high stability against oxidizing agents. This protects the sensor and the measuring point from drift and inaccurate measurements.
Thanks to Memosens technology, the sensors provide reliable, highly accurate measurements with a fast response time.
The inductive Memosens coupling between the corresponding Memosens sensor and the mobile CML18 handheld device is insensitive to moisture and corrosion.
The Memosens technology is based on a true plug-and-play principle. Just connect the sensor and you are ready to start your measurements. Measured values, calibration data and history are stored directly in the sensor. This means status values can be read out and a sensor check is also possible. An excellent feature to boost your GLP.
All sensors are supplied pre-calibrated. Thus, they are directly ready for use for your random sample measurement when the corresponding sensor is connected to the associated mobile handheld device Liquiline Mobile CML18.
 : EC_00828_03.22; EU_01000_01.22; UK_00482_01.25. Nonhazardous area approval, CE + UKCA marking.
Compatible handheld