Order Entry
Austria
ContactUsLinkComponent
 
pH Sensor Robust, Memosens CPL51E
  662-2458
undefined
pH Sensor Robust, Memosens CPL51E
  662-2458

 

  • Type:
    Potentiometric pH glass sensor with plastic shaft
  • Description:
    Memosens CPL51E pH sensor robust, 120 mm; open junction, 3 M KCl, Ag/AgCl ref. system
  • pH range:
    0 - 14
  • Connection:
    Memosens
  • Range:
    0 - 14 [application range 0 - 14 pH, 0…80 °C, 0,8…1,2 bar (abs.)]
  • Sensor type:
    Basic version, zero point Eo=7,0 temperature sensor NTC 30k
  • Reference system:
    Open junction, 3 M KCl, Ag/AgCl
  • Pk:
    1
  • pH:
    0 - 14 pH, 0…80 °C

 

 

Potentiometric pH glass sensor with plastic shaft for laboratory measurement and random sampling in the process. Connectable via inductive, digital connection with Memosens technology.

  • pH sensor for drinking water and wastewater applications
  • Polymer shaft protects the sensor from breaking
  • Open junction to minimize the risk of clogging

The Memosens 2.0 pH sensor CPL51E with a robust plastic shaft can be used both for analyzing process samples directly in the process or in the laboratory.

Thanks to Memosens technology, the sensors provide reliable, highly accurate measurements with a fast response time.

The inductive Memosens coupling between the corresponding Memosens sensor and the mobile CML18 handheld device is insensitive to moisture and corrosion.

The Memosens technology is based on a true plug-and-play principle. Just connect the sensor and you are ready to start your measurements. Measured values, calibration data and history are stored directly in the sensor. This means status values can be read out and a sensor check is also possible. An excellent feature to boost your GLP.

All sensors are supplied pre-calibrated. Thus, they are directly ready for use for your random sample measurement when the corresponding sensor is connected to the associated mobile handheld device Liquiline Mobile CML18.

 : EC_00828_03.22; EU_01000_01.22; UK_00482_01.22. Nonhazardous area approval, CE + UKCA marking.

Compatible handheld