BASE Footwear Yellowstone Safety Ankle Boots, Lace-UP, Mid, S3
: Portwest
Yellowstone mid safety boots feature a Microfibre + Hi-Tech fabric upper, SmellStop anti-bacterial lining, Fresh’n Flex midsole, SlimCap toe cap, and AirTech + TPU-skin sole.
- 200 joules resistant toe-cap
- Puncture-resistant sole
- Shock absorption in the heel region
- Anti-static footwear
- Electronic devices protection from electrostatic discharges
- Hydrocarbons-resistant
- Metal-free
Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces. Orthopaedic made to measure footbed, complying with european norms DGUV112-191.
: EN ISO 20345: 2011 S3 FO ESD SR; EN 61340-4-3 S3.
Frequently Bought Together