Order Entry
Austria
ContactUsLinkComponent
B-Phycoerythrin (BPE)
B-Phycoerythrin (BPE)
Catalog # COBSD4-000
CAS Number:  
undefined
B-Phycoerythrin (BPE)
Catalog # COBSD4-000
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Description:
    B-Phycoerythrin (B-PE), with SureLight® B-Phycoerythrin (B-PE) dye
  • Storage temperature:
    Store at 4 °C
  • Application:
    Flow cytometry, cell-based assays, microarrays and microplate applications
  • Ex/Em:
    545/572 nm

Specifications

About this item

Because of its high quantum yield, B-PE is considered the world’s brightest fluorophore.

It is compatible with commonly available lasers and gives exceptional results in flow cytometry, Luminex® and immunofluorescent staining. B-PE is also less 'sticky' than common synthetic fluorophores and therefore gives less background interference.