Orders LinkContactUsLinkComponent
Accessories for Modules
Accessories for Modules
Supplier:   SenseAnywhere
Accessories for Modules
Supplier:  SenseAnywhere
Certificates
Accessories for Modules
Accessories for Modules
5 Options Available Below
Product Details & Documents
Various Accessories for SenseAnywhere products.
  • Magnetic holders
  • Weather instrument shelter
  • Module connector lock clips
  • Protective cap
SenseAnywhere's Magnetic Holders are the perfect solution for attaching loggers to metal racks during Mapping in warehouses. The Magnetic Holders are designed with thermal insulation, ensuring the logger hangs slightly from the wall without impacting the temperature. This makes them an ideal solution for efficient Mapping in warehouses, where accuracy and precision are critical. Designed with convenience in mind, the magnets from the Magnetic Holder can also be easily attached to a Mobile AccessPoint, providing flexibility and ease when hanging it in a warehouse. Don't let the hassle of finding suitable mounting options for your loggers get in the way of efficient Mapping. Trust SenseAnywhere's Magnetic Holders to securely attach your loggers and get the job done with ease.
Magnetic Holders – The logger bracket can be attached to the Magnetic Holder using tape on the bracket. There are 3 mounting options for the Magnetic Holder. Magnetic: The magnets from the Magnetic Holder can be easily attached to metal racks. Tie wrap: The Magnetic Holder can be fastened to the desired surface using a Tie wrap. Hanging: The Magnetic Holder features a hole that allows a string to be threaded through, enabling it to be suspended in the air.
Weather Instrument Shelter – an accessory to protect your temperature and humidity measurements from unwanted weather influences such as rain or solar radiation. With the ability to easily mount to poles or other objects, the Weather Instrument Shelter is a versatile product that can be used in a variety of settings. The discs are made of durable UV resistant material and the stainless-steel mounting bracket guarantees a long service life. Investing in the Weather Instrument Shelter ensures that your temperature and humidity sensors remain accurate and reliable, providing you with valuable data that you can use to make informed decisions. Whether you need to monitor conditions in agriculture, construction, or environmental monitoring, the Weather Instrument Shelter is the perfect solution.
Module Connector Lock Clips – If you are using the SenseAnywhere Modules (e.g. Pt100 probe, 4 to 20 mA input module), the Lock Clip is available. This handy accessory keeps the connector securely in place so you can get on with your work without disruptions.
Protective Cap – Say goodbye to worries about dust and dirt getting into the AiroSensor 20-20-43 and 20-20-41 connector with our new Protective Cap. It keeps your connectors safe, so you can focus on what really matters.
Product Family Options
Product Information
  • Availability
    Price
  • SENA0096647495118
    Module connector lock clips
    Module connector lock clips
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 9,50 €
    9,50 €
    1 * 5 ST
       9,50 €
  • SENA0096647495125
    Module connector protective cap
    Module connector protective cap
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,00 €
    8,00 €
    1 * 1 ST
       8,00 €
  • SENA0096647495071
    Magnetic holder, small, 69 mm (for AiroSensor 20-20-2x and 20-20-3x), 69×33×3 mm; 0,028 kg; pull force magnets >10 kg
    Magnetic holder, small, 69 mm (for AiroSensor 20-20-2x and 20-20-3x), 69×33×3 mm; 0,028 kg; pull force magnets >10 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,60 €
    7,60 €
    1 * 1 ST
       7,60 €
  • SENA0096647495088
    Magnetic holder, large, 80 mm (for AiroSensor 20-20-4x and SwitchHolder), 80×34×3 mm; 0,032 kg; pull force magnets >10 kg
    Magnetic holder, large, 80 mm (for AiroSensor 20-20-4x and SwitchHolder), 80×34×3 mm; 0,032 kg; pull force magnets >10 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,60 €
    8,60 €
    1 * 1 ST
       8,60 €
  • SENA0096647495170
    Weather instrument shelter, 44 mm int.Ø, 140 mm ext.Ø., 200 mm H, 315 mm height with bracket, 175 mm depth from the post; 12 layers, around 147 mm (quantity and size), white with mirror finish, ABS engineering plastic (shelter material), stainless steel bracket, –40...+70 °C temperature range; 1,02 kg gross weight; (perfectly suitable for an AiroSensor with antenna connected to the 01-01-20 or 01-01-30 module)
    Weather instrument shelter, 44 mm int.Ø, 140 mm ext.Ø., 200 mm H, 315 mm height with bracket, 175 mm depth from the post; 12 layers, around 147 mm (quantity and size), white with mirror finish, ABS engineering plastic (shelter material), stainless steel bracket, –40...+70 °C temperature range; 1,02 kg gross weight; (perfectly suitable for an AiroSensor with antenna connected to the 01-01-20 or 01-01-30 module)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,80 €
    94,80 €
    1 * 1 ST
       94,80 €