Order Entry
Austria
ContactUsLinkComponent
 
CM Sephadex™ C-50 Ion Exchange Chromatography Media, Cytiva
 :  Cytiva
 
undefined
CM Sephadex™ C-50 Ion Exchange Chromatography Media, Cytiva
 :  Cytiva

 

  • pH range:
    6 - 10
  • Matrix:
    Cross-linked dextran
  • Ligand:
    Carboxymethyl
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    lgG (Mr 160 000)- 7 mg/ml drained medium), Bovine COHb (Mr 69 000)- 140 mg/ml drained medium), Ribonuclease (Mr 13 700)- 120 mg/ml drained medium)
  • Total ionic capacity:
    4 - 5 mmol/g
  • Description:
    CM Sephadex™ C-50

 

Frequently Bought Together