Orders LinkContactUsLinkComponent
High-speed barcode reader, VisionMate™ HSX
High-speed barcode reader, VisionMate™ HSX
High-speed barcode reader, VisionMate™ HSX
Catalog # 479-2448
   Low Manufacturing Impact
High-speed barcode reader, VisionMate™ HSX
Catalog # 479-2448
   Low Manufacturing Impact
High-speed barcode reader, VisionMate™ HSX
High-speed barcode reader, VisionMate™ HSX
High-speed barcode reader, VisionMate™ HSX
Price
11 850,00 €
Sale
Valid until 30/06/2026
Log in for availability
1 * 1 ST
Product Details & Documents
The Thermo Scientific™ VisionMate™ HSX High Speed Barcode Reader decodes 2D tube barcodes in as little as one second. Compatible with Matrix™ and Nunc™ tubes in 24, 48, 96, and 384 format racks.
  • Pre-loaded rack configurations allow you to rapidly read Matrix™ and Nunc™ racks of tubes without additional set up steps
  • Can decode both 2D and linear rack barcodes, providing additional tracking options
  • Customizable LED visual feedback provides the user with instant notification on successful reads, as well as notification of any read errors
  • Included magnetic linear barcode reader can be positioned in various locations on the all metal housing of the base unit to read linear rack barcodes.
  • Cyroprotection provides resistance to the accumulation of frost on the reading surface
  • Base unit is fully powered by a single USB connection to your computer
  • Can be used on the benchtop or integrated into your automated system
Supported Barcode Types: All Thermo Fisher Matrix™ and Nunc™ 2D Barcoded Tubes, ECC200, ISO 16022:2006, Linear Barcodes
The redesigned VisionMate™ Suite software provides a range of export options for seamless transfers of barcode data to your LIMS, ELN, or database.
The VisionMate™ HSX carries an IP66 rating, which protects the instrument from various adverse environmental conditions in your laboratory and establishes the critical environment barrier needed for use in a cleanroom.
A perfect complement to the Thermo Scientific™ Matrix™ and Nunc™ 2D barcoded tube offerings, the VisionMate™ HSX provides valuable process flexibility and compatibility in storage workflows in drug discovery, biological, or genomic storage environments.
Specifications
  • Description:
    High-speed barcode reader, VisionMate™ HSX
  • Interfaces:
    USB 2.0+
  • Cat. No.:
    479-2448
  • Environmental certification:
    NAR Certificate of Retirement
  • Memory:
    No internal storage capacity; data is stored on the user's computer
  • W×D×H:
    200×150×160 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Power:
    USB Powered