Orders LinkContactUsLinkComponent
Micro Test Tubes, Graduated
Micro Test Tubes, Graduated
Supplier:   KARTELL
Micro Test Tubes, Graduated
Supplier:  KARTELL
Certificates
Micro Test Tubes, Graduated
Micro Test Tubes, Graduated
4 Options Available Below
Product Details & Documents
Ideal for samples analysis.
  • Made from PP
  • Flat cap with writing area
  • Available in blue, yellow, pink and green
Depending on centrifuge being used, these tubes can withstand a centrifugal force of up to RCF 11000.
Certifications: The raw materials with the relevant requirements of Food-contact EU; Regulation 1935/2004 EC, Regulation 2023/2006/EC, FDA and REACH/ROHS regulations.
Product Family Options
Product Information
  • Capacity
    Colour
    Material
    Availability
    Price
  • KART0027904
    Micro test tube, graduated
    Micro test tube, graduated
    1,5 ml
    Blue
    PP
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 299,00 €
    299,00 €
    1 * 10.000 ST
       299,00 €
  • KART0027906
    Micro test tube, graduated
    Micro test tube, graduated
    1,5 ml
    Yellow
    PP
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 299,00 €
    299,00 €
    1 * 10.000 ST
       299,00 €
  • KART0027912
    Micro test tube, graduated
    Micro test tube, graduated
    1,5 ml
    Green
    PP
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 299,00 €
    299,00 €
    1 * 10.000 ST
       299,00 €
  • KART0027913
    Micro test tube, graduated
    Micro test tube, graduated
    1,5 ml
    Pink
    PP
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 299,00 €
    299,00 €
    1 * 10.000 ST
       299,00 €