Order Entry
Austria
ContactUsLinkComponent
SPC® Absorbents SitePoint Mobile Unit, Empty
SPC® Absorbents SitePoint Mobile Unit, Empty
Catalog # BRDY309075
Supplier:  Brady
undefined
SPC® Absorbents SitePoint Mobile Unit, Empty
Catalog # BRDY309075
Supplier:  Brady

Specifications

  • Description:
    Spill workstations SitePoint, mobile unit, empty
  • W×D×H:
    755,0×59,5×1725,0 mm
  • Absorbency:
    100 L
  • Pk:
    1

Specifications

About this item

Spill kit.

  • Features two, 200 mm wheels for mobility, and anchor points for fixed installation
  • Highly customisable
  • Fix items to backboard using self-tapping screws or bolts
  • Suitable for inside or outdoor use
  • Fully recyclable and chemical resistant

Ideal to move spill control equipment near specific risks. Features two rubber wheels for easy trolley movement to wherever it is required. Perfect during maintenance activities, at construction sites and other temporary locations. Includes three anchor points on the base, allowing the unit to be bolted to the ground, to avoid trolley movement without permission.