Core Inkjet Water Dissolvable Paper Roll Labels
: Brady
This B-403 material completely dissolves in warm water within 30 seconds, leaving no adhesive residue behind.
- Ideal for use in laboratory settings, asset tracking, or general use
- An easy to remove, environmentally responsible, print-on-demand inkjet label solution
- Easy removal saves time during relabeling process
- Pressure-sensitive adhesive allows for quick and easy application
- Designed for roll inkjet printers
- Matte finish
Applications include asset tracking, general identification, and laboratory labelling.
Frequently Bought Together