Elma Mutebox facilitates optimum cleaning, every time. It is hygienic, low-noise and environmentally friendly.
- Noise of ultrasonic cleaning unit operated in the Mutebox is reduced by up to 85%
- Vapors are exhausted via a silenced drain, so no heat or moisture builds up inside the box
- Laterally arranged halogen lamps
- Stainless steel jacket
Ingeniously innovative, Elma Mutebox guarantees low-noise cleaning. Its quiet fan and optimal insulation make working with ultrasonic equipment a pleasant, health-friendly and quiet experience, whether for cleaning or sample preparation. In addition, the fan extracts condensation water and vapours to the rear. Noise levels are reduced by up to 85% with the Elma Mutebox (ie. by a further 30% compared to the previous model).
The interior lighting and glass door allow the user to keep an eye on the cleaning process at all times. Two independent access points ensure that the items being cleaned and the ultrasonic unit are always easily accessible. The Mutebox, is made of high-quality steel and is easy to clean, thus also ensuring hygienic operation.
- Catalog No:
- ELMA1111901-001
- ELMA1123006
- W×D×H:
- Ext.: 545×530×525 mm; Int.: 430×380×445 mm
- Ext.: 860×605×525 mm; Int.: 710×450×445 mm
- Weight:
- 26,2 kg