ContactUsLinkComponent
Mop frame, Swep Duo Plus
Mop frame, Swep Duo Plus
Catalog # 129-0457
Supplier:  FHP VILEDA SCS
Mop frame, Swep Duo Plus
Catalog # 129-0457
Supplier:  FHP VILEDA SCS
Specifications
  • Description:
    Swep Duo Mop frame
  • Length:
    50 cm
  • Seq:
    06
  • Pk:
    1
Specifications
About this item
The double sided Swep Duo mop frame lets you clean twice the area by just flipping the frame and keep mopping. Swep Duo Plus frame with built-in lever is designed for touchless removal of dirty and/or contaminated mops.
  • Lightweight and ergonomic
  • Trapezoidal shape for easy cleaning of corners
  • Works with all core and specialist application double and single sided mops
Recommended handle: Aluminium telescopic handle (119966).