Quanti-Cult Plus™ Streptococcus pyogenes ATCC™ 19615™
Catalog # OXOIR4717000
Supplier: THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
- Pk:1 KIT
- Description:Streptococcus pyogenes ATCC™ 19615™
Specifications
About this item
Quanti-Cult Plus™ Streptococcus pyogenes ATCC™ 19615™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
10 preserved organism vials and 10 rehydration vials