Quanti-Cult Plus™ Staphylococcus aureus ATCC™ 6538P™
Catalog # OXOIR4719002
Supplier: THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
- Pk:1
- Description:Staphylococcus aureus ATCC™ 6538P™
Specifications
About this item
Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 6538P™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
According to ATCC S. aureus ATCC™ 6538P™ was found moderately resistant to penicillin in 1944 using a Penicillin P2 disk. This result was confirmed in initial characterization at ATCC, but subsequent investigation using a Penicillin P10 disk did not show resistance. Resistance has not been confirmed using modern methods in accordance with the Clinical & Laboratory Standards Institute guidelines.
Packaging: Includes 10 preserved organism vials and 10 rehydration vials.