Culti-Loops™ Schaalia odontolyticus ATCC™ 17929™
Catalog # OXOIR4600500
Supplier: THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
- Pk:5
- Description:Schaalia odontolyticus ATCC™ 17929™
- Seq:02
Specifications
About this item
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.