Quanti-Cult Plus™ Salmonella sp. serovar Abony NCTC 6017™
Catalog # OXOIR4716007
Supplier: THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
- Pk:1 KIT
- Description:Salmonella sp. serovar Abony NCTC 6017™
Specifications
About this item
Quanti-Cult Plus is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
Packaging: Includes 10 preserved organism vials and 10 rehydration vials.