ContactUsLinkComponent
Quanti-Cult Plus™ Pseudomonas aeruginosa ATCC™ 9027™
Quanti-Cult Plus™ Pseudomonas aeruginosa ATCC™ 9027™
  OXOIR4715210
 :  
undefined
Quanti-Cult Plus™ Pseudomonas aeruginosa ATCC™ 9027™
  OXOIR4715210
 :  
  • Pk:
    1 KIT
  • Description:
    Pseudomonas aeruginosa ATCC™ 9027™
  • Environmentally Preferable:
Quanti-Cult Plus™ Pseudomonas aeruginosa ATCC™ 9027™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
 : Includes 10 preserved organism vials and 10 rehydration vials.