ContactUsLinkComponent
Quanti-Cult Plus™ Candida albicans ATCC™ 2091™
Quanti-Cult Plus™ Candida albicans ATCC™ 2091™
Catalog # OXOIR4711601
CAS Number:  
undefined
Quanti-Cult Plus™ Candida albicans ATCC™ 2091™
Catalog # OXOIR4711601
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Candida albicans ATCC™ 2091™
  • Environmentally Preferable:
Specifications
About this item
Quanti-Cult Plus™ Candida albicans ATCC™ 2091™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1 ml inoculum
  • Ready for rehydration
  • For use in quality control procedures
Packaging: Includes 10 preserved organism vials and 10 rehydration vials.