ContactUsLinkComponent
Quanti-Cult Plus™ Burkholderia cepacia ATCC™ 25416™
Quanti-Cult Plus™ Burkholderia cepacia ATCC™ 25416™
Catalog # OXOIR4715220
CAS Number:  
undefined
Quanti-Cult Plus™ Burkholderia cepacia ATCC™ 25416™
Catalog # OXOIR4715220
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Burkholderia cepacia ATCC™ 25416™
Specifications
About this item
Quanti-Cult Plus™ Burkholderia cepacia ATCC™ 25416™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1 ml inoculum
  • Ready for rehydration
  • For use in quality control procedures
Packaging: Includes 10 preserved organism vials and 10 rehydration vials.