ContactUsLinkComponent
Quanti-Cult Plus™ Brevundimonas diminuta ATCC™ 19146™
Quanti-Cult Plus™ Brevundimonas diminuta ATCC™ 19146™
Catalog # OXOIR4711300
CAS Number:  
undefined
Quanti-Cult Plus™ Brevundimonas diminuta ATCC™ 19146™
Catalog # OXOIR4711300
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Brevundimonas diminuta ATCC™ 19146™
Specifications
About this item
Quanti-Cult Plus™ Brevundimonas diminuta ATCC™ 19146™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1 ml inoculum
  • 10 tests/reconstituted vial
  • 100 tests/kit
  • Ready for rehydration
  • For use in quality control procedures
Packaging: Includes 10 preserved organism vials and 10 rehydration vials.