Culti-Loops™ Aspergillus flavus ATCC™ 204304™
OXOIR4601312
:
- Pk:5
- Description:Aspergillus flavus ATCC™ 204304™
- Seq:59
Culti-Loops™ QC organisms. Aspergillus flavus ATCC™ 204304™ is recommended to be used for antifungal susceptibility testing quality control. The strain is recommended for antifungal MIC applications QC testing by CLSI and EUCAST.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.