ContactUsLinkComponent
Culti-Loops™ Alcaligenes faecalis subsp. faecalis ATCC® 8750™
Culti-Loops™ Alcaligenes faecalis subsp. faecalis ATCC® 8750™
Catalog # OXOIR4601006
CAS Number:  
undefined
Culti-Loops™ Alcaligenes faecalis subsp. faecalis ATCC® 8750™
Catalog # OXOIR4601006
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    Alcaligenes faecalis subsp. faecalis ATCC® 8750™
  • Seq:
    08
Specifications
About this item
Culti-Loops™ Alcaligenes faecalis subsp. faecalis ATCC™ 8750™ in the performance testing of culture media, stains, diagnostic kits, and reagents, for the maintenance of stock cultures and in the evaluation of bacteriological procedures. Culti-Loops™ are ready to use, disposable bacteriological loops containing stabilised viable microorganisms.
  • Ready-to-use - Prepared disposable bacteriological loops with viable microorganisms
  • Easy-to-store - Store at 2 to 8 °C
The film in each loop is made from a gelatine formulation and then dried by special processing. To rehydrate the film, the loops must come into contact with both warmth and moisture. Most organisms grow in 24 to 48 hours under the proper conditions. However, some exhibit a considerable lag phase and should be incubated for an additional 24 hours.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.