ContactUsLinkComponent
Culti-Loops™ Acinetobacter lwoff ATCC™ 17925™
Culti-Loops™ Acinetobacter lwoff ATCC™ 17925™
  OXOIR4601003
 :  
undefined
Culti-Loops™ Acinetobacter lwoff ATCC™ 17925™
  OXOIR4601003
 :  
  • Pk:
    5
  • Description:
    Acinetobacter lwoff ATCC™ 17925™
  • Seq:
    06
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
 : Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.