Easy-Tear Stopper Bags, with Steam Indicator, 1073B Tyvek/HDPE
Supplier: KEYSTONE CLEANROOM PRODUCTS
Certificates
About this item
Easy-Tear Tyvek bags from Keystone Cleanroom Products are designed to satisfy the most stringent requirements in the sterilisation of critical pharmaceutical equipment and components.
- Easy-Tear Feature for easier opening and sealing
- No sterile cutting instruments required to open the bag
- Low particle generation when opening via the Easy-Tear
- Manufactured and packaged in a certified ISO 6 cleanroom
- Exceptional microbial barrier
- Also compatible with gamma irradiation, V₂H₂O and EtO sterilisation
Delivers outstanding performance in terms of cleanliness, material and seal strength, as well as excellent microbial barrier properties.
Delivery information: Certificates of conformance sent with all shipments.
Packaging: Easy tear stopper bags are cleanroom packed in sealed, labeled poly bags with the Lot number and Date of manufacture.
Frequently Bought Together