ContactUsLinkComponent
Thermo Scientific™ Solaris 4000 I Large Incubated Benchtop Orbital Shakers
Thermo Scientific™ Solaris 4000 I Large Incubated Benchtop Orbital Shakers
Certificates
undefined
Thermo Scientific™ Solaris 4000 I Large Incubated Benchtop Orbital Shakers
Limited Time Sale
Select items below are currently on sale
Specifications
  • Catalog No:
  • 171-1519
  • 444-1438
  • BARNSK4001MPKG
  • Model:
  • Solaris 4000 I
  • Solaris 4000 I
  • Solaris 4000 I
  • Shaking motion:
  • Orbital
  • Orbital
  • Orbital
  • Temperature range:
  • 30…60 °C
  • 30…60 °C
  • 30…60 °C
  • Speed range:
  • 15 - 525 min⁻¹
  • 15 - 525 min⁻¹
  • 15 - 525 min⁻¹
  • Max. load:
  • 32 kg
  • 32 kg
  • 32 kg
  • Warranty:
  • Standard warranty includes 2 years labour, 5 years parts, and 10 years on the drive mechanism
  • Standard warranty includes 2 years labour, 5 years parts, and 10 years on the drive mechanism
  • Standard warranty includes 2 years labour, 5 years parts, and 10 years on the drive mechanism
  • Orbit:
  • 25 mm
  • 25 mm
  • 25 mm
  • Electrical:
  • 100 - 240 V, 50/60 Hz
  • 100 - 240 V, 50/60 Hz
  • 100 - 240 V, 50/60 Hz
  • Display:
  • Touch screen
  • Touch screen
  • Touch screen