Culti-Loops™ Candida dubliniensis ATCC™ MYA-577™
Catalog # OXOIR4609463
Supplier: THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
- Pk:5
- Description:Candida dubliniensis ATCC™ MYA-577™
Specifications
About this item
Culti-Loops™ Candida dubliniensis ATCC® 33560™ is recommended to be used for antimicrobial susceptibility quality control testing.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.