ContactUsLinkComponent
Culti-Loops™ Campylobacter jejuni ATCC® 33560™
Culti-Loops™ Campylobacter jejuni ATCC® 33560™
Catalog # OXOIR4609498
CAS Number:  
undefined
Culti-Loops™ Campylobacter jejuni ATCC® 33560™
Catalog # OXOIR4609498
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    Campylobacter jejuni ATCC® 33560™
Specifications
About this item
Confidently carry out testing of media, reagents, and identification and antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Culti-Loops™ QC organisms.
The loop may be dissolved in a liquid medium. Alternatively, it can be streaked directly onto appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.