ContactUsLinkComponent
Culti-Loops™ Staphylococcus epidermidis ATCC™ 29887™
Culti-Loops™ Staphylococcus epidermidis ATCC™ 29887™
Catalog # OXOIR4608209
CAS Number:  
undefined
Culti-Loops™ Staphylococcus epidermidis ATCC™ 29887™
Catalog # OXOIR4608209
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    Staphylococcus epidermidis ATCC™ 29887™
Specifications
About this item
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.