ContactUsLinkComponent
Cole-Parmer® ST-200 and SHP-200 Series 3-Position Magnetic Stirrers
Cole-Parmer® ST-200 and SHP-200 Series 3-Position Magnetic Stirrers
Supplier:  Cole Parmer
Certificates
undefined
Cole-Parmer® ST-200 and SHP-200 Series 3-Position Magnetic Stirrers
Supplier:  Cole Parmer
Specifications
  • Catalog No:
  • STUA51400-02
  • STUA51600-08
  • Model:
  • SHP-200-MP
  • No. of stirring positions:
  • 3
  • 3
  • Speed range:
  • 0 - 1500 min⁻¹
  • 100 - 1500 min⁻¹
  • Plate dimensions:
  • 120×550 mm
  • 160×160 mm
  • W×D×H:
  • 270×600×120 mm
  • 270×600×110 mm
  • Electrical:
  • 230 VAC, 50 Hz
  • 230 VAC, 50 Hz