Order Entry
Austria
Orders LinkContactUsLinkComponent
Aluminium acetate basic

Aluminium acetate basic

18 kg
Catalog # SPCMA1700-18KGBL
Supplier:   Spectrum Chemical
Aluminium acetate basic
Catalog # SPCMA1700-18KGBL
Supplier:  Spectrum Chemical
CAS Number:  142-03-0
Aluminium acetate basic

18 kg

CAS Number:  142-03-0
Price
6 310,00 €
Log in for availability
1 * 18 kg
Product Details & Documents

Aluminum Acetate Monobasic is a salt that is used as a topical astringent to treat skin irritations from poison ivy, substances such as soaps and cosmetics and insect bites. Aluminum acetate is particularly helpful for foot problems including athlete’s foot, swelling, and excessive sweating. It is also used to treat ear canal infections. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Synonyms:  Aluminium Acetate Monobasic
  • CAS Number:142-03-0
  • Formula:C₄H₇AlO₅
  • MDL Number:MFCD00008688
  • Storage Temperature:Ambient
  • EINECS:205-518-2

Documents

Specifications

Specification Test Results

  • Assay (%):
    28

Specifications

  • Pk:
    18 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.