Tyloxapol USP
Supplier: Spectrum Chemical
Jump to:
About this item
All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
- CAS Number:25301-02-4
- MDL Number:MFCD00149002
- Density:1,100
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
- CAS Number:25301-02-4
- MDL Number:MFCD00149002
- Density:1,100
- Storage Temperature:Ambient
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



