Dehydrocholic acid ≥98.5% USP
:81-23-2
:
:C₂₄H₃₄O₅
:MFCD00066410
:402.531 g/mol
:Ambient
- Assay (Dried Basis):98.5 - 101.0%
- Melting Range (Max. 3 C Range):231 - 242 °C
- Specific Rotation [A]D:+29.0 to +32.5
- Microbial Limits:To Pass Test
- Loss On Drying:max. 1.0 %
Frequently Bought Together