Hydroxypropyl methylcellulose (Hypromellose), 100,000 mPa.s USP Substitution Type 2208
Catalog # SPCMH1136-2.5KG
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Poly pail
- Pk:2,5 kg
- Seq:4076
- Environmentally Preferable:
Specifications
Specification Test Results
- Viscosity of a 2% Aqueous Solution @ 20D °C:75,000 - 140,000
- Assay (Methoxy Groups):19.0 - 24.0 %
- Assay (Hydroxypropoxy Groups):4.0 - 12.0 %
- pH:5.0 - 8.0
- Loss On Drying:max. 5.0 %
Specification Test Results
About this item
Synonyms: Methocel K100, Hydroxypropyl methyl cellulose
:
Storage Temperature:Ambient
CAS Number:9004-65-3
Formula:C₃H₈O₂·xCH₄O
MDL Number:MFCD00131360